Skip to Content
Merck
All Photos(1)

Key Documents

204886

Sigma-Aldrich

Complement C4, Human

Complement C4, Human, CAS 80295-48-3,is a native glycoprotein composed of three non-identical subunits of 93, 75, and 32 kDa linked by disulfide bonds.

Synonym(s):

Complement C4, Human

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.25

Quality Level

Assay

≥90% (SDS-PAGE)

form

liquid

specific activity

>3X105 C4H50 units/mg protein

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

shipped in

dry ice

storage temp.

−70°C

General description

Native, human C4 complement component. Glycoprotein composed of three non-identical subunits of M.W. 93,000 (α), 75,000 (β), and 32,000 (γ) linked by disulfide bonds. Present in normal human serum at 400 µg/ml. On activation of complement via the classical pathway, the C1s subcomponent of the C1 complex is converted to an active serine protease that cleaves the C4 α-chain at peptide bond 77, resulting in the production of C4a (M.W. 8740) and C4b fragments (M.W. 193,000). The released C4a peptide is one of the three complement-derived anaphylatoxins. The nascent C4b fragment can form a covalent ester bond with target surfaces. This covalent attachment of C4b to target acceptors is required for continuation of activation via classical pathway.
Native, human C4 complement component. Glycoprotein composed of three non-identical subunits of M.W. of 93 kDa (α), 75 kDa (β), and 32 kDa (γ) linked by disulfide bonds. Present in normal human serum at 400 µg/ml. On activation of complement via the classical pathway, the C1s subcomponent of the C1 complex is converted to an active serine protease that cleaves the C4 α-chain at peptide bond 77, resulting in production of C4a (M.W. 8740) and C4b fragments (M.W. 193 kDa). The released C4a peptide is one of the three complement-derived anaphylatoxins. The nascent C4b fragment can form a covalent ester bond with target surfaces. This covalent attachment of C4b to target acceptors is required for continuation of activation via classical pathway.

Biochem/physiol Actions

≥60% of the C4 activity present in freshly drawn normal human serum on a mg to mg basis

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Other Notes

Janatova, J. 1983. Ann. N.Y. Acad. Sci. 421, 218.
Hammer, C.H., et al. 1981. J. Biol. Chem. 256, 3995.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service