Skip to Content
Merck
All Photos(1)

Key Documents

1.93629

Supelco

Copper(II) nitrate trihydrate

EMPLURA®

Synonym(s):

Copper(II) nitrate trihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cu(NO3)2 · 3 H2O
CAS Number:
Molecular Weight:
241.60
MDL number:
UNSPSC Code:
12352302

grade

laboratory grade

product line

EMPLURA®

form

solid

density

2.05 g/cm3 at at 20 °C

storage temp.

room temp

InChI

1S/Cu.2NO3/c;2*2-1(3)4/q+2;2*-1

InChI key

XTVVROIMIGLXTD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Packaging

1.93629.0521: 500G, Minimum order quantity: 20

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Ox. Sol. 2 - Skin Corr. 1B

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Inorganics and solvents for basic and standard applications manufactured locally.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service