Skip to Content
Merck
All Photos(1)

Documents

05-778

Sigma-Aldrich

Anti-Rho (-A Antibody, -B, -C), clone 55

clone 55, Upstate®, from mouse

Synonym(s):

Aplysia ras-related homolog 12, oncogene RHO H12, ras homolog gene family, member A, small GTP binding protein RhoA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

55, monoclonal

species reactivity

human, mouse, rat

manufacturer/tradename

Upstate®

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... RHOA(387)

General description

Rho proteins belong to the Ras super family of low molecular weight GTPases, which includes RAS, RHO, RAB, RAN, RAD and ARF. These small GTP binding proteins function as molecular switches that regulate a multitude of biological processes including cell proliferation, apoptosis, differentiation, cytoskeletal reorganization, and membrane trafficking.

Specificity

Recognizes Rho (-A, -B, -C), Mr 22 kDa.

Immunogen

Recombinant human RhoA containing amino acids 1-155. Clone 55.

Application

Detect Rho (-A, -B, -C) using this Anti-Rho (-A Antibody, -B, -C), clone 55 validated for use in IC & WB.
Immunocytochemistry:
This antibody has been reported by an independent laboratory to detect Rho in cells via immunofluorescence.
Research Category
Signaling
Research Sub Category
Cytoskeletal Signaling

Quality

Routinely evaluated by western blot on RIPA lysates from Jurkat or HL-60 cells.

Western Blot Analysis:
0.5-2 µg/mL of this lot detected Rho in RIPA lysates from HL-60 cells. A previous lot detected Rho in RIPA lysates from Jurkat cells.

Target description

22 kDa

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1 in buffer containing BSA containing 0.09% sodium azide and 50% glycerol.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.

Handling Recommendations:
Upon receipt, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20ºC. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance. Note: Variability in freezer temperatures below -20ºC may cause glycerol-containing solutions to become frozen during storage.

Analysis Note

Control
HL-60 cell lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marina Sycheva et al.
Brain sciences, 9(8) (2019-08-23)
We have previously shown that the expression of pro-nerve growth factor (proNGF) was significantly increased, nerve growth factor (NGF) level was decreased, and the expression of p75NTR was enhanced in Alzheimer's disease (AD) hippocampal samples. NGF regulates cell survival and
Peroxynitrite mediates diabetes-induced endothelial dysfunction: possible role of Rho kinase activation.
El-Remessy, Azza B, et al.
Experimental Diabetes Research, 2010, 247861-247861 (2010)
Atorvastatin inhibits ABCA1 expression and cholesterol efflux in THP-1 macrophages by an LXR-dependent pathway.
Guosong Qiu,John S Hill
Journal of Cardiovascular Pharmacology null
Regulation of cell function by Rho GTPases.
DerMardirossian, C and Bokoch, G M
Drug News & Perspectives, 14, 389-395 (2001)
Myrto Raftopoulou et al.
Developmental biology, 265(1), 23-32 (2003-12-31)
Rho GTPases control signal transduction pathways that link cell surface receptors to a variety of intracellular responses. They are best known as regulators of the actin cytoskeleton, but in addition they control cell polarity, gene expression, microtubule dynamics and vesicular

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service