T-038
(+)-11-Nor-Δ9-THC-9-carboxylic acid glucuronide solution
100 μg/mL in methanol, ampule of 1 mL, Cerilliant®
About This Item
Recommended Products
grade
certified reference material
Quality Level
form
liquid
feature
Snap-N-Spike®/Snap-N-Shoot®
packaging
ampule of 1 mL
manufacturer/tradename
Cerilliant®
concentration
100 μg/mL in methanol
technique(s)
gas chromatography (GC): suitable
liquid chromatography (LC): suitable
application(s)
cannabis testing
forensics and toxicology
format
single component solution
storage temp.
−20°C
SMILES string
CCCCCc1cc(O)c2C3C=C(CCC3C(C)(C)Oc2c1)C(=O)OC4OC(C(O)C(O)C4O)C(O)=O
InChI
1S/C27H36O10/c1-4-5-6-7-13-10-17(28)19-15-12-14(8-9-16(15)27(2,3)37-18(19)11-13)25(34)36-26-22(31)20(29)21(30)23(35-26)24(32)33/h10-12,15-16,20-23,26,28-31H,4-9H2,1-3H3,(H,32,33)
InChI key
UWXZAZRSQQLWKH-UHFFFAOYSA-N
General description
Application
- Development of an automated sample preparation method based on solid phase extraction (SPE) to determine ∆9-tetrahydrocannabinol (THC) and its three primary metabolites in a variety of matrices, such as serum, saliva, and urine using liquid chromatography-mass spectrometry (LC-MS)
- Liquid chromatographic-tandem mass spectrometric (LC-MS/MS) determination of cannabinoids and their metabolites in whole blood samples
- Detection and quantification of 11-Nor-∆9-tetrahydrocannabinol-9-carboxylic acid glucuronide (THC-COOH-glu) in hair samples by ultra-performance liquid chromatography-tandem mass spectrometry (UHPLC-MS/MS)
- Simultaneous residue analysis of phytocannabinoids, their metabolites, and amphetamine-type stimulants (ATS) in 11 hair samples obtained from drug users by liquid chromatography-high resolution mass spectrometry (LC-HRMS)
- Determination of measurement uncertainty in the quantitative analysis of 11-nor-∆9-tetrahydrocannabinol-9-carboxylic acid (THC-COOH) and its glucuronide analogue (THC-COOH-glu) from urine samples by using LC-MS/MS
Features and Benefits
- Fully characterized under ISO/IEC 17025 and ISO 17034 accreditation
- Accompanied with a comprehensive Certificate of Analysis (CoA) with data on stability, homogeneity, accuracy of concentration, uncertainty, and traceability
- Rigorously tested through real-time stability studies to ensure accuracy and shelf life
- Gravimetrically prepared using qualified precision balances to ensure minimal uncertainty
- Flame sealed under argon into ampoules for long-term shelf life
- Offered in a convenient, DEA-exempt format to improve laboratory efficiency
Other Notes
Legal Information
related product
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1
Target Organs
Eyes
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
49.5 °F - closed cup
Flash Point(C)
9.7 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Protocols
Optimize β-glucuronidase hydrolysis for glucuronide metabolite analysis considering factors like time, temperature, pH, and enzyme concentration.
Optimize β-glucuronidase hydrolysis for glucuronide metabolite analysis considering factors like time, temperature, pH, and enzyme concentration.
Optimize β-glucuronidase hydrolysis for glucuronide metabolite analysis considering factors like time, temperature, pH, and enzyme concentration.
Optimize β-glucuronidase hydrolysis for glucuronide metabolite analysis considering factors like time, temperature, pH, and enzyme concentration.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service