W377406
Theaspirane
mixture of cis and trans, ≥85%
Synonym(s):
(±)-Theaspirane, 2,6,10,10-Tetramethyl-1-oxaspiro[4.5]dec-6-ene
About This Item
Kosher
Recommended Products
biological source
synthetic
Quality Level
grade
Halal
Kosher
reg. compliance
EU Regulation 178/2002
Assay
≥85%
refractive index
n20/D 1.438 (lit.)
bp
68-72 °C/3 mmHg (lit.)
density
0.931 g/mL at 25 °C (lit.)
application(s)
flavors and fragrances
Documentation
see Safety & Documentation for available documents
food allergen
no known allergens
Organoleptic
green; cooling; woody; herbaceous; camphoraceous
SMILES string
CC1CCC2(O1)C(C)=CCCC2(C)C
InChI
1S/C13H22O/c1-10-6-5-8-12(3,4)13(10)9-7-11(2)14-13/h6,11H,5,7-9H2,1-4H3
InChI key
GYUZHTWCNKINPY-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
- "Age" of tea: The impact of long-term storage on the aroma of Tuo tea and age prediction.: This study investigates the changes in the aroma profile of Tuo tea over long-term storage, identifying theaspirane as a key volatile compound. Theaspirane′s stability and impact on tea aroma make it significant for understanding aging processes and enhancing tea quality control methods (Chen et al., 2024).
Biochem/physiol Actions
Other Notes
Disclaimer
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
149.0 °F - closed cup
Flash Point(C)
65 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service