Key Documents
SYNV130501
Synthware conical bottom reaction vial with holed compression cap
0.1 mL, joint: ST/NS 5/5
Synonym(s):
Synthware V130501, reaction tube
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
Pricing and availability is not currently available.
Joint
joint: ST/NS5/5
packaging
pkg of 1 ea
manufacturer/tradename
Synthware V130501
capacity
0.1 mL
Related Categories
General description
Supplied with one holed compression cap, one Viton® O-ring and one PTFE -faced silicone rubber septum. For replacement caps, see RC112; For Viton® O-rings, see R014; for septa, see R1420.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)