Key Documents
SYNA752440
Synthware 90 degree vacuum adapter with glass stopcock
male joint: ST/NS 24/40
Synonym(s):
Synthware A752440, flow control adapter
About This Item
UNSPSC Code:
41121815
NACRES:
NB.51
Pricing and availability is not currently available.
Joint
male joint: ST/NS24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware A752440
General description
Used to regulate the flow of gases or liquids into reaction systems. Stopcock has a bore of 2 mm.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)