Skip to Content
Merck
All Photos(1)

Key Documents

932264

Sigma-Aldrich

Poly[(9,9-dioctylfluorenyl-2,7-diyl)-alt-(6,6′-{2,2′-bipyridine})]

Mw ≥10,000 Da by GPC

Synonym(s):

PFO-BPy

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C39H46N2)n
CAS Number:
UNSPSC Code:
12352207
NACRES:
NA.23

mol wt

Mw ≥10,000 Da by GPC

Quality Level

energy gap

3.4 eV (Eg (HOMO-LUMO gap))

solubility

THF: soluble
chloroform: soluble

λmax

360 nm±5 nm in THF

fluorescence

λem 373 nm±10 nm in THF
λem 392 nm±10 nm in THF

Orbital energy

HOMO 6.2 eV 
LUMO 2.8 eV 

Application

Polymer Emitting Layers for Solution-Processed OLEDs.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High-Yield Dispersions of Large-Diameter Semiconducting Single-Walled Carbon Nanotubes with Tunable Narrow Chirality Distributions
Mistry, Kevin S., et al.
ACS Nano, 7, 2231-2239 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service