Skip to Content
Merck
All Photos(1)

Key Documents

564494

Sigma-Aldrich

4-Fluoro-2-methoxyphenylboronic acid

≥95%

Synonym(s):

4-Fluoro-2-methoxybenzeneboronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H3(OCH3)B(OH)2
CAS Number:
Molecular Weight:
169.95
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95%

form

powder

mp

131-136 °C (lit.)

SMILES string

COc1cc(F)ccc1B(O)O

InChI

1S/C7H8BFO3/c1-12-7-4-5(9)2-3-6(7)8(10)11/h2-4,10-11H,1H3

InChI key

ADJBXDCXYMCCAD-UHFFFAOYSA-N

Application

Reactant involved in:
  • Suzuki-Miyaura cross-coupling reactions for synthesis of heterobiaryls
  • Suzuki and Still coupling for synthesis of antithrombotic drugs

Reactant / precursor used for the synthesis of biologically active molecules including:
  • 2-(4-pyridyl)thienopyridinones for use as GSK-3β inhibitors
  • Arylheteroarylmethylamines as NR2B subtype of NMDA receptor antagonists with antidepressant activity
  • Biphenylketones for use as dual modulators of inflammation and bone loss
  • CB1 antagonists

Other Notes

Contains varying amounts of anhydride

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service