Skip to Content
Merck
All Photos(1)

Key Documents

496812

Sigma-Aldrich

7-Allyl-7,8-dihydro-8-oxoguanosine

95%

Synonym(s):

Loxoribine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H17N5O6
CAS Number:
Molecular Weight:
339.30
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

optical activity

[α]21/D −32°, c = 1 in H2O

mp

227-230 °C (lit.)

SMILES string

NC1=NC2=C(N(CC=C)C(=O)N2[C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O)C(=O)N1

InChI

1S/C13H17N5O6/c1-2-3-17-6-9(15-12(14)16-10(6)22)18(13(17)23)11-8(21)7(20)5(4-19)24-11/h2,5,7-8,11,19-21H,1,3-4H2,(H3,14,15,16,22)/t5-,7-,8-,11-/m1/s1

InChI key

VDCRFBBZFHHYGT-IOSLPCCCSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Loxoribine is a selective toll-like receptor 7 (TLR7) agonist that activates immune cells and increases cytokine production.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marion Jurk et al.
European journal of immunology, 36(7), 1815-1826 (2006-06-20)
Toll-like receptors (TLR) 7 and 8 are closely related members of the TLR family of pathogen-associated molecular pattern recognition receptors and have an important function in activation of innate immune responses upon viral infection. TLR7 can be activated selectively by
Kristie A Jenkins et al.
Developmental and comparative immunology, 33(5), 660-667 (2008-12-23)
Although Toll-like receptors (TLRs) have been well characterised in mammals, less work has been carried out in non-mammalian species, such as chickens. In this study the response of chicken cells to the TLR9 subfamily of ligands was characterised in vitro
Mouldy Sioud et al.
European journal of immunology, 37(10), 2834-2846 (2007-09-14)
We recently reported that human bone marrow hematopoietic CD34(+) progenitors express functional Toll-like receptors (TLR) and can differentiate into myeloid cells just by stimulation with resiquimod (R848), a specific agonist for TLR7/8. However, the mechanisms by which R848 induces cell
C L Swaggerty et al.
Poultry science, 91(4), 1038-1042 (2012-03-09)
Young poultry exhibit a transient colonization by some food-borne pathogens, including Salmonella, during the first week of life that stems from immature innate and acquired defense mechanisms. Consequently, modulation of the hosts' natural immune response is emerging as an important
Saradiya Chatterjee et al.
Cancer research, 74(18), 5008-5018 (2014-07-31)
Toll-like receptors (TLR) recognize pathogen molecules and danger-associated signals that stimulate inflammatory processes. TLRs have been studied mainly in antigen-presenting cells, where they exert important immune regulatory functions, but they are also expressed by epithelial tumor cells, where they have

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service