Skip to Content
Merck
All Photos(1)

Key Documents

489379

Sigma-Aldrich

Ethylenediaminetetraacetic-d12 acid

98 atom % D

Synonym(s):

EDTA-d12

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HO2CCD2)2N(CD2)2N(CD2CO2H)2
CAS Number:
Molecular Weight:
304.32
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

98 atom % D

form

solid

technique(s)

NMR: suitable
bio NMR: suitable

mp

250 °C (dec.) (lit.)

mass shift

M+12

SMILES string

[2H]C([2H])(N(C([2H])([2H])C(O)=O)C([2H])([2H])C([2H])([2H])N(C([2H])([2H])C(O)=O)C([2H])([2H])C(O)=O)C(O)=O

InChI

1S/C10H16N2O8/c13-7(14)3-11(4-8(15)16)1-2-12(5-9(17)18)6-10(19)20/h1-6H2,(H,13,14)(H,15,16)(H,17,18)(H,19,20)/i1D2,2D2,3D2,4D2,5D2,6D2

InChI key

KCXVZYZYPLLWCC-LBTWDOQPSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mallika Sastry et al.
Journal of biomolecular NMR, 50(3), 197-207 (2011-06-15)
NMR spectroscopic characterization of the structure or the dynamics of proteins generally requires the production of samples isotopically enriched in (15)N, (13)C, or (2)H. The bacterial expression systems currently in use to obtain isotopic enrichment, however, cannot produce a number
Dan Li et al.
Journal of proteome research, 14(5), 2237-2254 (2015-03-19)
Atherosclerosis resulting from hyperlipidemia causes many serious cardiovascular diseases. To understand the systems changes associated with pathogenesis and progression of atherosclerosis, we comprehensively analyzed the dynamic metabonomic changes in multiple biological matrices of LDLR(-/-) mice using NMR and GC-FID/MS with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service