Skip to Content
Merck
All Photos(2)

Key Documents

265063

Sigma-Aldrich

Beryllium

chunks, 99.9%

Synonym(s):

Glucinium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Be
CAS Number:
Molecular Weight:
9.01
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99.9%

form

chunks

autoignition temp.

1198 °F

reaction suitability

reagent type: Lewis acid
core: beryllium

reagent type: catalyst

resistivity

4.46 μΩ-cm, 20°C

bp

2970 °C (lit.)

mp

1278 °C (lit.)

density

1.85 g/mL at 25 °C (lit.)

SMILES string

[Be]

InChI

1S/Be

InChI key

ATBAMAFKBVZNFJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Starting material for novel sterically encumbered arylberyllium compounds.

Reagent for:
  • Synthesis of beryllium phthalocyanine
  • Metal mediated dechlorination

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Carc. 1B - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R. Kubiak, et al.,
Inorgorganica Chimica Acta, 359, 1344-1350 (2006)
Shengli Zhu et al.
Journal of the mechanical behavior of biomedical materials, 13, 166-173 (2012-08-18)
We developed Ni- and Be-free Zr(45+x)Cu(40-x)Al₇Pd₅Nb₃ bulk metallic glasses with large glass-forming ability and unusual plasticity. The alloys have large critical diameters (larger than 10 mm) in a wide composition range (x=0-20). The Zr₅₀Cu₃₅Al₇Pd₅Nb₃ and Zr₅₅Cu₃₀Al₇Pd₅Nb₃ alloys exhibit the largest
Mark Niemeyer et al.
Inorganic chemistry, 36(21), 4688-4696 (2001-10-24)
The use of the terphenyl substituent -C(6)H(3)-2,6-Mes(2) (abbreviated Ar) has permitted the synthesis of several new low-coordinate beryllium compounds. Reaction of 1 equiv of LiAr with BeCl(2)(OEt(2))(2) or BeBr(2)(OEt(2))(2) (1) gives the monomeric complexes ArBeX(OEt(2)) (X = Cl (2); Br
A Hermanne et al.
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine, 70(12), 2763-2772 (2012-10-10)
New experimental data for production of (7)Be and (22,24)Na in deuteron irradiation of (nat)Mg and Al up to 50 MeV are presented. The induced activity, measured with HPGe spectroscopy, allows us to determine excitation functions of (nat)Mg(d,x) and (27)Al(d,x) reactions
V. K. Tandon and R. B. Chhor,
Letters in Organic Chemistry, 1, 40-42 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service