Skip to Content
Merck
All Photos(1)

Documents

225185

Sigma-Aldrich

Cyclobutylamine

98%

Synonym(s):

Aminocyclobutane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C4H7NH2
CAS Number:
Molecular Weight:
71.12
Beilstein:
2069297
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.437 (lit.)

bp

81.5 °C/752 mmHg (lit.)

density

0.833 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

NC1CCC1

InChI

1S/C4H9N/c5-4-2-1-3-4/h4H,1-3,5H2

InChI key

KZZKOVLJUKWSKX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

24.8 °F - closed cup

Flash Point(C)

-4 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hartmut Schirok
The Journal of organic chemistry, 71(15), 5538-5545 (2006-07-15)
7-Azaindoles are versatile building blocks, especially in medicinal chemistry, where they serve as bioisosteres of indoles or purines. Herein, we are presenting a robust and flexible synthesis of 1,3- and 1,3,6-substituted 7-azaindoles starting from nicotinic acid derivatives or 2,6-dichloropyridine, respectively.
T Maruyama et al.
Chemical & pharmaceutical bulletin, 38(10), 2719-2725 (1990-10-01)
9-Cyclobutyladenine (4a), cis- and trans-9-[3- (hydroxymethyl)cyclobutyl]adenine (4b) and 9-[3,3-bis(hydroxymethyl)cyclobutyl]adenine(4d) were prepared from the corresponding cyclobutylamine derivatives (1a, 1b and 1d). Guanine congeners (9a, cis- and trans-9b and 9d) and carbocyclic oxetanocin G (1',2'-trans-9f) were also prepared. Carbocyclic oxetanocin A(1',2'-trans-4f), the
McNaughton et al.
Journal of molecular spectroscopy, 196(2), 274-282 (1999-11-30)
The infrared spectrum of vinylamine, generated by pyrolysis of cyclobutylamine, has been investigated at low and high resolution. The rovibrational structure of the far infrared spectrum (0.002 cm(-1)) has been analyzed, and effective rotational and centrifugal distortion constants have been

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service