HPLC Analysis of Furocoumarins on Ascentis® Express RP-Amide
CONDITIONS
column
Ascentis Express RP-Amide, 10 cm x 4.6 mm I.D., 2.7 μm particles (53929-U)
mobile phase
[A] water; [B] methanol; (40:60, A:B)
flow rate
1.0 mL/min
pressure
2843 psi (196 bar)
column temp.
35 °C
detector
UV, 254 nm
injection
5 μL
sample
25 μg/mL in water:methanol (90:10)
Description
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany