Merck
  • Selective F or Br Functionalization of Dibenzofuran for Application as Host Materials of Phosphorescent Organic Light-Emitting Diodes.
Selective F or Br Functionalization of Dibenzofuran for Application as Host Materials of Phosphorescent Organic Light-Emitting Diodes.
Chemistry (Weinheim an der Bergstrasse, Germany) (2017-08-24)
Ji Gwang Yu, Sung Yong Byeon, Si Hyun Han, Jun Yeob Lee
ABSTRACT
Four dibenzofuran-type host materials substituted with a carbazolylcarbazole moiety were synthesized to investigate the effect of substitution position on the material parameters and device performances of host materials. The carbazolylcarbazole moiety was substituted at the 1-, 2-, 3-, and 4-positions of dibenzofuran by F or Br for a comprehensive study of the positional effect of dibenzofuran-derived host materials. Systematic synthesis and comparison of the four host materials revealed that 1-, 2-, and 4-position modification was better than 3-position modification for high triplet energy and high external quantum efficiency.
Sigma-Aldrich
2-Methoxyphenylboronic acid, 95%
Sigma-Aldrich
1-Bromo-2,6-difluorobenzene, 98%
Sigma-Aldrich
1,10-Phenanthroline, ≥99%