Merck
  • The Hippo signaling functions through the Notch signaling to regulate intrahepatic bile duct development in mammals.
The Hippo signaling functions through the Notch signaling to regulate intrahepatic bile duct development in mammals.
Laboratory investigation; a journal of technical methods and pathology (2017-06-06)
Nan Wu, Quy Nguyen, Ying Wan, Tiaohao Zhou, Julie Venter, Gabriel A Frampton, Sharon DeMorrow, Duojia Pan, Fanyin Meng, Shannon Glaser, Gianfranco Alpini, Haibo Bai
ABSTRACT
The Hippo signaling pathway and the Notch signaling pathway are evolutionary conserved signaling cascades that have important roles in embryonic development of many organs. In murine liver, disruption of either pathway impairs intrahepatic bile duct development. Recent studies suggested that the Notch signaling receptor Notch2 is a direct transcriptional target of the Hippo signaling pathway effector YAP, and the Notch signaling is a major mediator of the Hippo signaling in maintaining biliary cell characteristics in adult mice. However, it remains to be determined whether the Hippo signaling pathway functions through the Notch signaling in intrahepatic bile duct development. We found that loss of the Hippo signaling pathway tumor suppressor Nf2 resulted in increased expression levels of the Notch signaling pathway receptor Notch2 in cholangiocytes but not in hepatocytes. When knocking down Notch2 on the background of Nf2 deficiency in mouse livers, the excessive bile duct development induced by Nf2 deficiency was suppressed by heterozygous and homozygous deletion of Notch2 in a dose-dependent manner. These results implicated that Notch signaling is one of the downstream effectors of the Hippo signaling pathway in regulating intrahepatic bile duct development.