Merck
  • Ethanol Extract of Potentilla supina Linne Suppresses LPS-induced Inflammatory Responses through NF-κB and AP-1 Inactivation in Macrophages and in Endotoxic mice.
Ethanol Extract of Potentilla supina Linne Suppresses LPS-induced Inflammatory Responses through NF-κB and AP-1 Inactivation in Macrophages and in Endotoxic mice.
Phytotherapy research : PTR (2017-01-28)
Hae-Jun Lee, Ji-Sun Shin, Kyoung-Goo Lee, Sang Cheol Park, Young Pyo Jang, Jung-Hwan Nam, Kyung-Tae Lee
ABSTRACT
In this study, we investigated the antiinflammatory effects of ethanol extracts of Potentilla. supina Linne (EPS) in lipopolysaccharide (LPS)-induced RAW 264.7 macrophages and septic mice. EPS suppressed LPS-induced nitric oxide, prostaglandin E
Sigma-Aldrich
NS-398, ≥98% (HPLC), solid
Sigma-Aldrich
Vanadium(III) chloride, 97%