Merck
3-
Experimental and therapeutic medicine (2017-10-01)
Juan Chen, Jingyang Wang, Jiyan Zhang, Chuanqiang Pu
ABSTRACT
3-
Sigma-Aldrich
Freund′s Adjuvant, Complete, cell suspension
Sigma-Aldrich
Myosin, Calcium activated from rabbit muscle, buffered aqueous glycerol solution, 0.5-1.5 units/mg protein (biuret)
Sigma-Aldrich
Pertussis toxin from Bordetella pertussis, lyophilized powder