Merck
  • Generation and intracellular trafficking of a polysialic acid-carrying fragment of the neural cell adhesion molecule NCAM to the cell nucleus.
Generation and intracellular trafficking of a polysialic acid-carrying fragment of the neural cell adhesion molecule NCAM to the cell nucleus.
Scientific reports (2017-08-19)
Nina Westphal, Gabriele Loers, David Lutz, Thomas Theis, Ralf Kleene, Melitta Schachner
ABSTRACT
Polysialic acid (PSA) and its major protein carrier, the neural cell adhesion molecule NCAM, play important roles in many nervous system functions during development and in adulthood. Here, we show that a PSA-carrying NCAM fragment is generated at the plasma membrane by matrix metalloproteases and transferred to the cell nucleus via endosomes and the cytoplasm. Generation and nuclear import of this fragment in cultured cerebellar neurons is induced by a function-triggering NCAM antibody and a peptide comprising the effector domain (ED) of myristoylated alanine-rich C kinase substrate (MARCKS) which interacts with PSA within the plane of the plasma membrane. These treatments lead to activation of the fibroblast growth factor (FGF) receptor, phospholipase C (PLC), protein kinase C (PKC) and phosphoinositide-3-kinase (PI3K), and subsequently to phosphorylation of MARCKS. Moreover, the NCAM antibody triggers calmodulin-dependent activation of nitric oxide synthase, nitric oxide (NO) production, NO-dependent S-nitrosylation of matrix metalloprotease 9 (MMP9) as well as activation of matrix metalloprotease 2 (MMP2) and MMP9, whereas the ED peptide activates phospholipase D (PLD) and MMP2, but not MMP9. These results indicate that the nuclear PSA-carrying NCAM fragment is generated by distinct and functionally defined signal transducing mechanisms.
Sigma-Aldrich
CGS-9343B, ≥98% (HPLC), powder
Sigma-Aldrich
FAK Inhibitor 14, ≥95% (HPLC)