Merck
Activation of CB
American journal of physiology. Regulatory, integrative and comparative physiology (2017-03-31)
Olga Karpińska, Marta Baranowska-Kuczko, Monika Kloza, Ewa Ambroz Ewicz, Tomasz Kozłowski, Irena Kasacka, Barbara Malinowska, Hanna Kozłowska
ABSTRACT
Recent evidence suggests that endocannabinoids acting via cannabinoid CB
Sigma-Aldrich
JZL 184 hydrate, ≥97% (HPLC)
Sigma-Aldrich
9,11-Dideoxy-11α,9α-epoxymethanoprostaglandin F, solution, 10 mg/mL in methyl acetate
Sigma-Aldrich
AM251, >98% (HPLC), solid
Supelco
Serotonin, analytical standard
Sigma-Aldrich
AM630, ≥90% (HPLC)