Merck
  • (tBu2MeSi)2Sn=Sn(SiMetBu2)2: a distannene with a >Sn=Sn< double bond that is stable both in the solid state and in solution.
(tBu2MeSi)2Sn=Sn(SiMetBu2)2: a distannene with a >Sn=Sn< double bond that is stable both in the solid state and in solution.
Journal of the American Chemical Society (2006-08-31)
Vladimir Ya Lee, Tomohide Fukawa, Masaaki Nakamoto, Akira Sekiguchi, Boris L Tumanskii, Miriam Karni, Yitzhak Apeloig
ABSTRACT
((t)Bu(2)MeSi)(2)Sn=Sn(SiMe(t)Bu(2))(2) 1, prepared by the reaction of (t)Bu(2)MeSiNa with SnCl(2)-diox in THF and isolated as dark-green crystals, represents the first example of acyclic distannene with a Sn=Sn double bond that is stable both in the crystalline form and in solution. This was proved by the crystal and NMR spectral data of 1. Distannene 1 has these peculiar structural features: a shortest among all acyclic distannenes Sn=Sn double bond of 2.6683(10) A, a nearly planar geometry around both Sn atoms, and a highly twisted Sn=Sn double bond. The reactions of 1 toward carbon tetrachloride and phenylacetylene also correspond to the reactivity anticipated for the Sn=Sn double bond. The one-electron reduction of 1 with potassium produced the distannene anion radical, the heavy analogue of alkene ion radicals, for which the particular crystal structure and low-temperature EPR behavior are also discussed.
Sigma-Aldrich
4,7,13,16,21,24-Hexaoxa-1,10-diazabicyclo[8.8.8]hexacosane, 98%
Pricing and availability is not currently available.