Merck
  • Increased expression of the PI3K catalytic subunit p110δ underlies elevated S6 phosphorylation and protein synthesis in an individual with autism from a multiplex family.
Increased expression of the PI3K catalytic subunit p110δ underlies elevated S6 phosphorylation and protein synthesis in an individual with autism from a multiplex family.
Molecular autism (2016-01-16)
Ashwini C Poopal, Lindsay M Schroeder, Paul S Horn, Gary J Bassell, Christina Gross
ABSTRACT
Dysfunctions in the PI3K/mTOR pathway have gained a lot of attention in autism research. This was initially based on the discovery of several monogenic autism spectrum disorders with mutations or defects in PI3K/mTOR signaling components. Recent genetic studies corroborate that defective PI3K/mTOR signaling might be a shared pathomechanism in autism disorders of so far unknown etiology, but functional molecular analyses in human cells are rare. The goals of this study were to perform a functional screen of cell lines from patients with idiopathic autism for defects in PI3K/mTOR signaling, to test if further functional analyses are suitable to detect underlying molecular mechanisms, and to evaluate this approach as a biomarker tool to identify therapeutic targets. We performed phospho-S6- and S6-specific ELISA experiments on 21 lymphoblastoid cell lines from the AGRE collection and on 37 lymphoblastoid cell lines from the Simons Simplex Collection and their healthy siblings. Cell lines from one individual with increased S6 phosphorylation and his multiplex family were analyzed in further detail to identify upstream defects in PI3K signaling associated with autism diagnosis. We detected significantly increased S6 phosphorylation in 3 of the 21 lymphoblastoid cell lines from AGRE compared to a healthy control and in 1 of the 37 lymphoblastoid cell lines from the Simons Simplex Collection compared to the healthy sibling. Further analysis of cells from one individual with elevated S6 phosphorylation showed increased expression of the PI3K catalytic subunit p110δ, which was also observed in lymphoblastoid cells from other autistic siblings but not unaffected members in his multiplex family. The p110δ-selective inhibitor IC87114 reduced elevated S6 phosphorylation and protein synthesis in this cell line. Our results suggest that functional analysis of PI3K/mTOR signaling is a biomarker tool to identify disease-associated molecular defects that could serve as therapeutic targets in autism. Using this approach, we discovered impaired signaling and protein synthesis through the PI3K catalytic subunit p110δ as an underlying molecular defect and potential treatment target in select autism spectrum disorders. Increased p110δ activity was recently associated with schizophrenia, and our results suggest that p110δ may also be implicated in autism.
Sigma-Aldrich
Anti-PI3 Kinase Antibody, p110β, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone B-5-1-2, purified from hybridoma cell culture