Merck
  • Effects of surface-active block copolymers with oxyethylene and fluoroalkyl side chains on the antifouling performance of silicone-based films.
Effects of surface-active block copolymers with oxyethylene and fluoroalkyl side chains on the antifouling performance of silicone-based films.
Biofouling (2016-01-16)
Elisa Martinelli, Deniz Gunes, Brandon M Wenning, Christopher K Ober, John A Finlay, Maureen E Callow, James A Callow, Alessio Di Fino, Anthony S Clare, Giancarlo Galli
ABSTRACT
Block copolymers made from a poly(dimethyl siloxane) (Si) and a poly(meth)acrylate carrying oxyethylene (EG) or fluoroalkyl (AF) side chains were synthesized and incorporated as surface-active components into a silicone matrix to produce cross-linked films with different surface hydrophilicity/phobicity. Near-edge X-ray absorption fine structure (NEXAFS) studies showed that film surfaces containing Si-EG were largely populated by the siloxane, with the oxyethylene chains present only to a minor extent. In contrast, the fluorinated block was selectively segregated to the polymer-air interface in films containing Si-AF as probed by NEXAFS and X-ray photoelectron spectroscopy (XPS) analyses. Such differences in surface composition were reflected in the biological performance of the coatings. While the films with Si-EG showed a higher removal of both Ulva linza sporelings and Balanus amphitrite juveniles than the silicone control, those with Si-AF exhibited excellent antifouling properties, preventing the settlement of cyprids of B. amphitrite.
Sigma-Aldrich
Bismuth neodecanoate
Sigma-Aldrich
Poly(dimethylsiloxane), viscosity 1.0 cSt (25 °C)
Sigma-Aldrich
3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluorooctyl acrylate, contains inhibitor, 97%
Sigma-Aldrich
1,1,4,7,10,10-Hexamethyltriethylenetetramine, 97%
Sigma-Aldrich
α-Bromoisobutyryl bromide, 98%