Merck
  • γ-Secretase directly sheds the survival receptor BCMA from plasma cells.
γ-Secretase directly sheds the survival receptor BCMA from plasma cells.
Nature communications (2015-06-13)
Sarah A Laurent, Franziska S Hoffmann, Peer-Hendrik Kuhn, Qingyu Cheng, Yuanyuan Chu, Marc Schmidt-Supprian, Stefanie M Hauck, Elisabeth Schuh, Markus Krumbholz, Heike Rübsamen, Johanna Wanngren, Mohsen Khademi, Tomas Olsson, Tobias Alexander, Falk Hiepe, Hans-Walter Pfister, Frank Weber, Dieter Jenne, Hartmut Wekerle, Reinhard Hohlfeld, Stefan F Lichtenthaler, Edgar Meinl
ABSTRACT
Survival of plasma cells is regulated by B-cell maturation antigen (BCMA), a membrane-bound receptor activated by its agonist ligands BAFF and APRIL. Here we report that γ-secretase directly cleaves BCMA, without prior truncation by another protease. This direct shedding is facilitated by the short length of BCMA's extracellular domain. In vitro, γ-secretase reduces BCMA-mediated NF-κB activation. In addition, γ-secretase releases soluble BCMA (sBCMA) that acts as a decoy neutralizing APRIL. In vivo, inhibition of γ-secretase enhances BCMA surface expression in plasma cells and increases their number in the bone marrow. Furthermore, in multiple sclerosis, sBCMA levels in spinal fluid are elevated and associated with intracerebral IgG production; in systemic lupus erythematosus, sBCMA levels in serum are elevated and correlate with disease activity. Together, shedding of BCMA by γ-secretase controls plasma cells in the bone marrow and yields a potential biomarker for B-cell involvement in human autoimmune diseases.
Sigma-Aldrich
Anti-BCMA antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Nicastrin antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
TAPI-1, TAPI-1, CAS 171235-71-5, is a structural analog of TAPI-0 with similar in vitro efficacy for the inhibition of MMPs and TACE. Blocks the shedding of several cell surface proteins.
Sigma-Aldrich
γ-Secretase Inhibitor IX, Gamma-Secretase Inhibitor IX - CAS 208255-80-5, is a cell-permeable inhibitor of γ-secretase (Aβtotal IC₅₀ = 115 nM, Aβ42 IC₅₀ = 200 nM).