Merck
  • Synthesis, Characterization, Antimicrobial and Antitumor Activities of Sucrose Octa(N-ethyl)carbamate.
Synthesis, Characterization, Antimicrobial and Antitumor Activities of Sucrose Octa(N-ethyl)carbamate.
Medicinal chemistry (Shariqah (United Arab Emirates)) (2015-08-11)
Claudia D Raposo, Krasimira T Petrova, Maria T Barros, Ricardo C Calhelha, Marina Sokovic, Isabel C F R Ferreira
ABSTRACT
Sucrose octa(N-ethyl)carbamate was synthesized directly from sucrose and ethyl isocyanate, and its structure was confirmed by various analytical methods, such as (1)H and (13)C NMR, FTIR, m.p., MS, and optical rotation. Its antibacterial, antifungal and cytotoxic activities were investigated. It exhibited strong inhibition against all bacteria tested, namely S. aureus (MIC 0.18±0.006), B. cereus (MIC 0.094±0.000), M. flavus (MIC 0.28±0.01), L. monocytogenes (MIC 0.18±0.006), P. aeruginosa (MIC 0.094±0.002), S. typhimurium (MIC 0.094±0.002), E. coli (MIC 0.18±0.006) and E. cloacae (MIC 0.18±0.006) and strong antifungal activity towards T. viride (MIC 0.09 ± 0.006), A. versicolor (MIC 0.18 ± 0.01), A. ochraceus (MIC 0.375 ± 0.01) and P. ochrochloron (MIC 0.375 ± 0.04). Furthermore, it showed moderate antitumor potential against human breast (GI50 357.20±14.12), colon (GI50 332.43±11.19) and cervical (GI50 282.67±3.97) cell lines and, more important, without hepatotoxicity.
Sigma-Aldrich
Ethyl isocyanate, 98%
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Ampicillin, meets USP testing specifications
Sigma-Aldrich
Triethylamine, ≥99%
HCT-15, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA, 91030712, human colon, Epithelial-like
Millipore
Bacillus cereus NCTC 7464 Lenticule® discs, 500-50000 CFU, suitable for microbiology, certified reference material, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Pyridine, ACS reagent, ≥99.0%
MCF7 Cell Line human, 86012803, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA, 86012803, human breast (adenocarcinoma), Epithelial-like
Millipore
Listeria monocytogenes NCTC 11994 Lenticule® discs, 500-50000 CFU, suitable for microbiology, certified reference material, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland