Merck
  • Accelerated separation of GC-amenable lipid classes in plant oils by countercurrent chromatography in the co-current mode.
Accelerated separation of GC-amenable lipid classes in plant oils by countercurrent chromatography in the co-current mode.
Analytical and bioanalytical chemistry (2015-10-07)
Simon Hammann, Michael Englert, Marco Müller, Walter Vetter
ABSTRACT
Triacylglycerols represent the major part (>90%) in most plant oils and have to be eliminated, when the minor compounds such as phytosterols or tocopherols should be analyzed. Here, we used an all liquid-liquid chromatographic technique, countercurrent chromatography (CCC), to fractionate the minor lipids before gas chromatography (GC) analysis. To cover the wide range of polarity of the minor compounds, we used the co-current mode, in which both mobile and stationary phase are pumped through the system. This allowed to elute substances which partitioned almost exclusively in the stationary phase within 90 min. After testing with standard compounds, the method was applied to the separation of sesame oil and sunflower oil samples. The abundant triacylglycerols could be effectively separated from tocopherols, phytosterols, diacylglycerols, and free fatty acids in the samples, and these compounds could be analyzed (after trimethylsilylation) by GC coupled with mass spectrometry. After the enrichment caused by the CCC fractionation, we were also able to identify the tocopherol derivative α-tocomonoenol, which had not been described in sunflower oil before. Also, separation of sesame oil yielded a mixture of the polar compounds sesamin and sesamolin without further impurities.
Product Description
Supelco
5-α-Cholestane, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
5-α-Cholestane, ≥97.0% (HPLC)
Sigma-Aldrich
Palmitic acid, ≥99%
Supelco
N,O-Bis(trimethylsilyl)trifluoroacetamide with trimethylchlorosilane, with 1% trimethylchlorosilane, for GC derivatization, LiChropur
Sigma-Aldrich
Palmitic acid, BioXtra, ≥99%
Sigma-Aldrich
Palmitic acid, ≥98% palmitic acid basis (GC)
Sigma-Aldrich
Aluminum oxide, mesoporous, MSU-X (wormhole), average pore size 3.8 nm
Sigma-Aldrich
2,3,4-Trifluorotoluene, 99%
Sigma-Aldrich
Palmitic acid, ≥98%, FCC, FG
Sigma-Aldrich
Chlorotrimethylsilane, produced by Wacker Chemie AG, Burghausen, Germany, ≥99.0% (GC)
Sigma-Aldrich
Chlorotrimethylsilane, ≥98.0% (GC)
Sigma-Aldrich
1,1,3,3-Tetramethyl-1,3-diphenyldisilazane, 96%
Sigma-Aldrich
Chlorotrimethylsilane, purified by redistillation, ≥99%
Sigma-Aldrich
Cholesteryl stearate, 96%
Sigma-Aldrich
Palmitic acid, natural, 98%, FG
Sigma-Aldrich
Pyridine, ≥99.5% (GC)
Sigma-Aldrich
α,α,α-Trifluorotoluene solution, NMR reference standard, 0.05% in benzene-d6 (99.6 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Aluminum oxide, nanowires, diam. × L 2-6 nm × 200-400 nm
Sigma-Aldrich
N,O-Bis(trimethylsilyl)trifluoroacetamide, ≥99%
Sigma-Aldrich
α,α,α-Trifluorotoluene solution, NMR reference standard, 0.05% in benzene-d6 (99.6 atom % D)
Sigma-Aldrich
Pyridine, ≥99%
Sigma-Aldrich
Methyl oleate, technical grade, 70%
Sigma-Aldrich
Chlorotrimethylsilane solution, 1.0 M in THF
Sigma-Aldrich
Methyl oleate, 99%
Sigma-Aldrich
Pyridine, anhydrous, 99.8%
Sigma-Aldrich
Aluminum oxide, 99.997% trace metals basis
Sigma-Aldrich
Oleic acid, technical grade, 90%
Sigma-Aldrich
Aluminum oxide, nanopowder, <50 nm particle size (TEM)
Sigma-Aldrich
Oleic acid, ≥99% (GC)
Sigma-Aldrich
Oleic acid, BioReagent, suitable for cell culture