Merck
  • SKLB-287, a novel oral multikinase inhibitor of EGFR and VEGFR2, exhibits potent antitumor activity in LoVo colorectal tumor model.
SKLB-287, a novel oral multikinase inhibitor of EGFR and VEGFR2, exhibits potent antitumor activity in LoVo colorectal tumor model.
Neoplasma (2014-07-18)
X Chen, Y Liu, H-W Yang, S Zhou, C Cheng, M-W Zheng, L Zhong, X-Y Fu, Y-L Pan, S Ma, Y Tang, Y-Z Chen, L-L Li, S-Y Yang
ABSTRACT
Colorectal cancer (CRC) is the third common cancer and most of the chemotherapies of CRC currently used often suffer limited efficacy and large side effects. Targeted small-molecule by anti-tumor drugs are thought a promising strategy for improving the efficacy and reducing the side effects. In this investigation, we report a novel multikinase inhibitor, termed SKLB-287, which was discovered by us recently. SKLB-287 could efficiently inhibit the activation of endothelial growth factor receptor (EGFR) and vascular endothelial growth factor receptor 2 (VEGFR2). It displayed very good anti-proliferative activity against LoVo CRC cells and considerable antiangiogenic potency in transgenic zebrafish embryos. Oral administration of SKLB-287 resulted in dose-dependent suppression of tumor growth in LoVo xenograft mouse model. Immunohistochemistry was adopted to examine the in vivo anti-tumor mechanism of action of SKLB-287.
Sigma-Aldrich
Anti-phospho-EGFR (pThr693) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%