Merck
  • Silver-zeolite combined to polyphenol-rich extracts of Ascophyllum nodosum: potential active role in prevention of periodontal diseases.
Silver-zeolite combined to polyphenol-rich extracts of Ascophyllum nodosum: potential active role in prevention of periodontal diseases.
PloS one (2014-10-02)
Zohreh Tamanai-Shacoori, Fatiha Chandad, Amélie Rébillard, Josiane Cillard, Martine Bonnaure-Mallet
ABSTRACT
The purpose of this study was to evaluate various biological effects of silver-zeolite and a polyphenol-rich extract of A. nodosum (ASCOP) to prevent and/or treat biofilm-related oral diseases. Porphyromonas gingivalis and Streptococcus gordonii contribute to the biofilm formation associated with chronic periodontitis. In this study, we evaluated in vitro antibacterial and anti-biofilm effects of silver-zeolite (Ag-zeolite) combined to ASCOP on P. gingivalis and S. gordonii growth and biofilm formation capacity. We also studied the anti-inflammatory and antioxidant capacities of ASCOP in cell culture models. While Ag-zeolite combined with ASCOP was ineffective against the growth of S. gordonii, it showed a strong bactericidal effect on P. gingivalis growth. Ag-zeolite combined with ASCOP was able to completely inhibit S. gordonii monospecies biofilm formation as well as to reduce the formation of a bi-species S. gordonii/P. gingivalis biofilm. ASCOP alone was ineffective towards the growth and/or biofilm formation of S. gordonii and P. gingivalis while it significantly reduced the secretion of inflammatory cytokines (TNFα and IL-6) by LPS-stimulated human like-macrophages. It also exhibited antioxidant properties and decreased LPS induced lipid peroxidation in gingival epithelial cells. These findings support promising use of these products in future preventive or therapeutic strategies against periodontal diseases.
Product Description
Supelco
Menadione (K3), analytical standard
Menadione, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Menadione, meets USP testing specifications
Sigma-Aldrich
Menadione, crystalline
USP
Menadione, United States Pharmacopeia (USP) Reference Standard
Supelco
Menadione, Pharmaceutical Secondary Standard; Certified Reference Material