Merck
  • Thiazide-sensitive Na+ -Cl- cotransporter (NCC) gene inactivation results in increased duodenal Ca2+ absorption, enhanced osteoblast differentiation and elevated bone mineral density.
Thiazide-sensitive Na+ -Cl- cotransporter (NCC) gene inactivation results in increased duodenal Ca2+ absorption, enhanced osteoblast differentiation and elevated bone mineral density.
Journal of bone and mineral research : the official journal of the American Society for Bone and Mineral Research (2014-07-06)
Yu-Juei Hsu, Sung-Sen Yang, Chih-Jen Cheng, Shu-Ting Liu, Shih-Ming Huang, Tom Chau, Pauling Chu, Donald M Salter, Herng-Sheng Lee, Shih-Hua Lin
ABSTRACT
Inactivation of the thiazide-sensitive sodium chloride cotransporter (NCC) due to genetic mutations in Gitelman's syndrome (GS) or pharmacological inhibition with thiazide diuretics causes hypocalciuria and increased bone mineral density (BMD) with unclear extrarenal calcium (Ca(2+) ) regulation. We investigated intestinal Ca(2+) absorption and bone Ca(2+) metabolism in nonsense Ncc Ser707X (S707X) homozygous knockin mice (Ncc(S707X/S707X) mice). Compared to wild-type and heterozygous knockin littermates, Ncc(S707X/S707X) mice had increased intestinal absorption of (45) Ca(2+) and expression of the active Ca(2+) transport machinery (transient receptor potential vanilloid 6, calbindin-D9K , and plasma membrane Ca(2+) ATPase isoform 1b). Ncc(S707X/S707X) mice had also significantly increased Ca(2+) content accompanied by greater mineral apposition rate (MAR) in their femurs and higher trabecular bone volume, cortical bone thickness, and BMD determined by μCT. Their osteoblast differentiation markers, such as bone alkaline phosphatase, procollagen I, osteocalcin, and osterix, were also significantly increased while osteoclast activity was unaffected. Analysis of marrow-derived bone cells, either treated with thiazide or directly cultured from Ncc S707X knockin mice, showed that the differentiation of osteoblasts was associated with increased phosphorylation of mechanical stress-induced focal adhesion kinase (FAK) and extracellular signal-regulated kinase (ERK). In conclusion, NCC inhibition stimulates duodenal Ca(2+) absorption as well as osteoblast differentiation and bone Ca(2+) storage, possibly through a FAK/ERK dependent mechanism.
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Supelco
Calcium standard for AAS, analytical standard, 1.000 g/L Ca+2 in hydrochloric acid, traceable to BAM
SAFC
HEPES
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
7-Aminoactinomycin D, ~97% (HPLC), powder
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
Nitrotetrazolium Blue chloride, ≥90.0% (HPLC)
Sigma-Aldrich
Nitrotetrazolium Blue chloride, powder, electrophoresis grade
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
6-Benzylaminopurine, suitable for plant cell culture
SAFC
HEPES
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
6-Benzylaminopurine, ReagentPlus®, ≥99.0% (HPLC)
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Supelco
L-Ascorbic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
6-Benzylaminopurine, PESTANAL®, analytical standard
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material