Merck
  • Dual-modified liposomes with a two-photon-sensitive cell penetrating peptide and NGR ligand for siRNA targeting delivery.
Dual-modified liposomes with a two-photon-sensitive cell penetrating peptide and NGR ligand for siRNA targeting delivery.
Biomaterials (2015-02-24)
Yang Yang, YanFang Yang, XiangYang Xie, ZhiYuan Wang, Wei Gong, Hui Zhang, Ying Li, FangLin Yu, ZhiPing Li, XingGuo Mei
ABSTRACT
Tumor-oriented nanocarrier drug delivery approaches with photosensitivity have drawn considerable attention over the years. However, due to its low penetrability and ability to harm tissues, the use of UV light for triggered nanocarrier release in in vivo applications has been limited. Compared with UV light, near-infrared (NIR) light deeply penetrates tissues and is less damaging to cells. In this study, we devised and tested a strategy for functional siRNA delivery to cells by loading siRNA into cationic liposomes bearing a photolabile-caged cell-penetrating peptide (pcCPP) and asparagine-glycine-arginine peptide (NGR) molecules attached to the liposome surface (pcCPP/NGR-LP). Here, the positive charges of the lysine residues on the CPP were temporarily caged by the photosensitive group (PG), neutralizing its charges and thereby forming a pcCPP. This event subsequently led to conditional NIR light-dependent cell-penetrating functionality. After administration, the pcCPP/NRG-LP was inactivated in the circulatory system as it could not penetrate the tumor cell membrane. The NGR moiety selectively bound to CD13-positive tumors, which facilitated the active accumulation of pcCPP/NGR-LP in tumor tissues. Then, upon illumination using NIR light at the tumor site, the PG was uncaged, the interaction of the CPP with the cell membrane was restored and the activated dual-modified liposomes exhibited enhanced tumor cellular uptake and selectivity due to the synergistic effect of CPP-mediated cellular entry and NGR-mediated endocytosis. Subsequent research demonstrated that the pcCPP/NGR-LP showed good physicochemical properties, effective cellular uptake, endosomal escape and significant gene silencing in HT-1080 cells in vitro. Additionally, after systemic administration in mice, pcCPP/NGR-LP accumulated in the tumor, augmented c-myc silencing and delayed tumor progression. In conclusion, the combined application of these pcCPP and NGR modifications may provide a reasonable approach for the selectively targeted delivery of siRNA.
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, aqueous solution, sterile-filtered, suitable for cell culture
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, purum, ≥99% (GC)
Sigma-Aldrich
Cholesterol, tested according to Ph. Eur.
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Sodium pyrophosphate tetrabasic, ≥95%
Sigma-Aldrich
Sodium orthovanadate, ≥90% (titration)
Sigma-Aldrich
Triethylamine, ≥99.5%
Supelco
Triethylamine, analytical standard
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Triethylamine, for protein sequence analysis, ampule, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, ≥99%
Sigma-Aldrich
Triethylamine, for amino acid analysis, ≥99.5% (GC)
Sigma-Aldrich
Sodium orthovanadate, 99.98% trace metals basis
Sigma-Aldrich
Cholesteryl N-(2-dimethylaminoethyl)carbamate, ≥98% (TLC)
Sigma-Aldrich
Cholesterol, from lanolin, ≥99.0% (GC)
Supelco
Cholesterol, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
Cholesterol, Plant-Derived, SyntheChol®
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Supelco
Dichloromethane, Selectophore, for ion-selective electrodes, ≥99.5%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Dichloromethane, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 50-150 ppm amylene as stabilizer
Supelco
Methylene Chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder