Merck
  • The responses of autophagy and apoptosis to oxidative stress in nucleus pulposus cells: implications for disc degeneration.
The responses of autophagy and apoptosis to oxidative stress in nucleus pulposus cells: implications for disc degeneration.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology (2014-10-04)
Jiang-Wei Chen, Bin-Bin Ni, Bo Li, Yue-Hua Yang, Sheng-Dan Jiang, Lei-Sheng Jiang
ABSTRACT
Apoptosis and autophagy are two patterns of programmed cell death which play important roles in the intervertebral disc degeneration. Oxidative stress is an important factor for the induction of programmed cell death. However, the cellular reactions linking autophagy to apoptosis of disc cells under oxidative stress have never been described. This study investigated the responses of autophagy and apoptosis and their interactions in the nucleus pulposus cells (NP cells) under oxidative stress, with the aim to better understand the mechanism of disc degeneration. NP cells isolated from rat lumbar discs were subjected to different concentrations of H2O2 for various time periods. Cell viability was determined by CCK-8 assay, and their apoptosis and autophagy responses were evaluated by fluorescent analysis, flow cytometry and western blotting, et al. The interactions of autophagy and apoptosis and the possible signaling pathways were also investigated by using autophagy modulators. H2O2 increased the lysosomal membrane permeability in the NP cells and induced apoptosis through the mitochondrial pathway subsequently. Meanwhile, H2O2 stimulated an early autophagy response through the ERK/m-TOR signaling pathway. Autophagy inhibition significantly decreased the apoptosis incidence in the cells insulted by H2O2. These results suggested that controlling the autophagy response in the NP cells under oxidative stress should be beneficial for the survival of the cells and probably delay the process of disc degeneration.
Sigma-Aldrich
3-Methyladenine, autophagy inhibitor
Sigma-Aldrich
Acridine, BioReagent, suitable for fluorescence, ≥97.0% (HPLC)
Sigma-Aldrich
SP600125, ≥98% (HPLC)
Sigma-Aldrich
Acridine, ≥96.5% (HPLC)