Merck
  • The differential characterization of GPR55 receptor in human peripheral blood reveals a distinctive expression in monocytes and NK cells and a proinflammatory role in these innate cells.
The differential characterization of GPR55 receptor in human peripheral blood reveals a distinctive expression in monocytes and NK cells and a proinflammatory role in these innate cells.
International immunology (2014-10-27)
Valerio Chiurchiù, Mirko Lanuti, Marco De Bardi, Luca Battistini, Mauro Maccarrone
ABSTRACT
G protein-coupled receptor 55 (GPR55) is activated by endogenous, plant-derived and synthetic cannabinoids. Recent studies reported a broad tissue distribution for GPR55 and found prominent roles for this receptor in inflammatory pain, gut and bone physiology, as well as cancer. However, little is known about the expression and function of GPR55 in immune cells. To address this question, we performed a detailed characterization of GPR55 in different human innate and adaptive immune populations using polychromatic flow cytometry and we found that monocytes and NK cells expressed remarkable levels of this receptor compared to several cells of adaptive immunity. GPR55 activation by the specific agonist O-1602 boosted IL-12 and TNF-α production, and decreased endocytic activity, in LPS-activated monocytes. In addition, it increased CD69 activation marker expression, granzyme B and CD107a-dependent cytotoxicity and IFN-γ and TNF-α production in NK cells activated by both IL-2 and IL-12. These over-stimulatory effects of GPR55 were antagonized by its selective antagonist cannabidiol. Altogether, our data thus unveil a proinflammatory role for GPR55 in innate immunity that may be important for the design of new immune therapeutic strategies.
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Resorcinol, ≥98%, FG
Sigma-Aldrich
Resorcinol, ReagentPlus®, 99%
Sigma-Aldrich
Resorcinol, meets analytical specification of Ph. Eur., BP, 98.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Resorcinol, ACS reagent, ≥99.0%
Sigma-Aldrich
Brefeldin A, ≥99% (HPLC and TLC), BioXtra, for molecular biology
Sigma-Aldrich
Brefeldin A, from Penicillium brefeldianum, ≥99% (HPLC and TLC)
Sigma-Aldrich
Brefeldin A, from Penicillium brefeldianum, Ready Made Solution, 10 mg/mL in DMSO
Supelco
Resorcinol, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Resorcinol, BioXtra, ≥99%
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Resorcinol, European Pharmacopoeia (EP) Reference Standard
Hymecromone impurity A, European Pharmacopoeia (EP) Reference Standard
USP
Resorcinol, United States Pharmacopeia (USP) Reference Standard