Merck
  • Neuropeptide receptors provide a signalling pathway for trigeminal modulation of olfactory transduction.
Neuropeptide receptors provide a signalling pathway for trigeminal modulation of olfactory transduction.
The European journal of neuroscience (2012-12-05)
Philipp Daiber, Federica Genovese, Valentin A Schriever, Thomas Hummel, Frank Möhrlen, Stephan Frings
ABSTRACT
The mammalian olfactory epithelium contains olfactory receptor neurons and trigeminal sensory endings. The former mediate odor detection, the latter the detection of irritants. The two apparently parallel chemosensory systems are in reality interdependent in various well-documented ways. Psychophysical studies have shown that virtually all odorants can act as irritants, and that most irritants have an odor. Thus, the sensory perception of odorants and irritants is based on simultaneous input from the two systems. Moreover, functional interactions between the olfactory system and the trigeminal system exist on both peripheral and central levels. Here we examine the impact of trigeminal stimulation on the odor response of olfactory receptor neurons. Using an odorant with low trigeminal potency (phenylethyl alcohol) and a non-odorous irritant (CO(2) ), we have explored this interaction in psychophysical experiments with human subjects and in electroolfactogram (EOG) recordings from rats. We have demonstrated that simultaneous activation of the trigeminal system attenuates the perception of odor intensity and distorts the EOG response. On the molecular level, we have identified a route for this cross-modal interaction. The neuropeptide calcitonin-gene related peptide (CGRP), which is released from trigeminal sensory fibres upon irritant stimulation, inhibits the odor response of olfactory receptor neurons. CGRP receptors expressed by these neurons mediate this neuromodulatory effect. This study demonstrates a site of trigeminal-olfactory interaction in the periphery. It reveals a pathway for trigeminal impact on olfactory signal processing that influences odor perception.
Sigma-Aldrich
cAMP Enzyme Immunoassay Kit, Direct
Sigma-Aldrich
Anti-Calcitonin Gene Related Peptide (CGRP) Rabbit pAb, lyophilized, Calbiochem®
Sigma-Aldrich
Anti-CRCP antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, ≥99% (HPLC), powder