Merck
  • A JNK-mediated autophagy pathway that triggers c-IAP degradation and necroptosis for anticancer chemotherapy.
A JNK-mediated autophagy pathway that triggers c-IAP degradation and necroptosis for anticancer chemotherapy.
Oncogene (2013-07-09)
W He, Q Wang, B Srinivasan, J Xu, M T Padilla, Z Li, X Wang, Y Liu, X Gou, H-M Shen, C Xing, Y Lin
ABSTRACT
Killing cancer cells through the induction of apoptosis is one of the main mechanisms of chemotherapy. However, numerous cancer cells have primary or acquired apoptosis resistance, resulting in chemoresistance. In this study, using a novel chalcone derivative chalcone-24 (Chal-24), we identified a novel anticancer mechanism through autophagy-mediated necroptosis (RIP1- and RIP3-dependent necrosis). Chal-24 potently killed different cancer cells with induction of necrotic cellular morphology while causing no detectable caspase activation. Blocking the necroptosis pathway with either necrostatin-1 or by knockdown of RIP1 and RIP3 effectively blocked the cytotoxicity of Chal-24, suggesting that Chal-24-induced cell death is associated with necroptosis. Chal-24 robustly activated JNK and ERK and blockage of which effectively suppressed Chal-24-induced cytotoxicity. In addition, Chal-24 strongly induced autophagy that is dependent on JNK-mediated phosphorylation of Bcl-2 and Bcl-xL and dissociation of Bcl-2 or Bcl-xL from Beclin-1. Importantly, suppression of autophagy, with either pharmacological inhibitors or small interfering RNAs targeting the essential autophagy components ATG7 and Beclin-1, effectively attenuated Chal-24-induced cell death. Furthermore, we found that autophagy activation resulted in c-IAP1 and c-IAP2 degradation and formation of the Ripoptosome that contributes to necroptosis. These results thus establish a novel mechanism for killing cancer cells that involves autophagy-mediated necroptosis, which may be employed for overcoming chemoresistance.
Sigma-Aldrich
MISSION® esiRNA, targeting human RALBP1
Sigma-Aldrich
MISSION® esiRNA, targeting human RIPK1
Sigma-Aldrich
MISSION® esiRNA, targeting human ATG7
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Ralbp1
Sigma-Aldrich
Adenosine 5′-diphosphoribose sodium salt, ≥93%
Sigma-Aldrich
Anti-LC3B antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
MISSION® esiRNA, targeting human UQCRFS1
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Atg7
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Ripk1
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
SP600125, ≥98% (HPLC)