Merck
  • Stromal heparan sulfate differentiates neuroblasts to suppress neuroblastoma growth.
Stromal heparan sulfate differentiates neuroblasts to suppress neuroblastoma growth.
The Journal of clinical investigation (2014-06-18)
Erik H Knelson, Angela L Gaviglio, Jasmine C Nee, Mark D Starr, Andrew B Nixon, Stephen G Marcus, Gerard C Blobe
ABSTRACT
Neuroblastoma prognosis is dependent on both the differentiation state and stromal content of the tumor. Neuroblastoma tumor stroma is thought to suppress neuroblast growth via release of soluble differentiating factors. Here, we identified critical growth-limiting components of the differentiating stroma secretome and designed a potential therapeutic strategy based on their central mechanism of action. We demonstrated that expression of heparan sulfate proteoglycans (HSPGs), including TβRIII, GPC1, GPC3, SDC3, and SDC4, is low in neuroblasts and high in the Schwannian stroma. Evaluation of neuroblastoma patient microarray data revealed an association between TGFBR3, GPC1, and SDC3 expression and improved prognosis. Treatment of neuroblastoma cell lines with soluble HSPGs promoted neuroblast differentiation via FGFR1 and ERK phosphorylation, leading to upregulation of the transcription factor inhibitor of DNA binding 1 (ID1). HSPGs also enhanced FGF2-dependent differentiation, and the anticoagulant heparin had a similar effect, leading to decreased neuroblast proliferation. Dissection of individual sulfation sites identified 2-O, 3-O-desulfated heparin (ODSH) as a differentiating agent, and treatment of orthotopic xenograft models with ODSH suppressed tumor growth and metastasis without anticoagulation. These studies support heparan sulfate signaling intermediates as prognostic and therapeutic neuroblastoma biomarkers and demonstrate that tumor stroma biology can inform the design of targeted molecular therapeutics.
Sigma-Aldrich
Citrate Concentrated Solution, BioUltra, for molecular biology, 1 M in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
Citrate Concentrated Solution, BioReagent, suitable for coagulation assays, 4 % (w/v)
Pricing and availability is not currently available.
Sigma-Aldrich
Heparan sulfate proteoglycan, ≥400 μg/mL glycosaminoglycan
Pricing and availability is not currently available.
Tretinoin, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
Supelco
Tretinoin, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
USP
Tretinoin, United States Pharmacopeia (USP) Reference Standard
Pricing and availability is not currently available.
Supelco
Sodium Citrate, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Sigma-Aldrich
Retinoic acid, ≥98% (HPLC), powder
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-FGF-2/basic FGF (neutralizing) Antibody, clone bFM-1, clone bFM-1, Upstate®, from mouse
Pricing and availability is not currently available.
Sigma-Aldrich
MISSION® esiRNA, targeting human SDC3
Pricing and availability is not currently available.
Sigma-Aldrich
Retinoic acid p-hydroxyanilide, ≥95%
Pricing and availability is not currently available.
Sigma-Aldrich
Heparin sodium salt from porcine intestinal mucosa, Grade I-A, ≥180 USP units/mg, powder, BioReagent, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
PD 173074, ≥96% (HPLC), powder
Pricing and availability is not currently available.