Merck
Acute accidental lindane ingestion in toddlers.
Annals of emergency medicine (1995-11-01)
S E Aks, A Krantz, D O Hryhrczuk, S Wagner, J Mock
ABSTRACT
Lindane toxicity has been reported to occur mostly by way of dermal exposure. Cases of ingestion in which blood levels have been determined are rare. We present three such cases, along with a comparison of cases in the literature with respect to blood level half-lives and correlation with signs of toxicity. Emergency physicians can prevent acute ingestion by educating patients on the proper use of lindane and by selecting less toxic scabicidal agents.
Product Description
Supelco
Lindane, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Lindane, PESTANAL®, analytical standard
Supelco
γ-BHC, certified reference material, 1000 μg/mL in methanol