Merck
  • Efficacy and safety of pegylated interferon alfa-2a or alfa-2b plus ribavirin for the treatment of chronic hepatitis C in children and adolescents: a systematic review and meta-analysis.
Efficacy and safety of pegylated interferon alfa-2a or alfa-2b plus ribavirin for the treatment of chronic hepatitis C in children and adolescents: a systematic review and meta-analysis.
Clinical infectious diseases : an official publication of the Infectious Diseases Society of America (2012-12-18)
Eric Druyts, Kristian Thorlund, Ping Wu, Steve Kanters, Sanni Yaya, Curtis L Cooper, Edward J Mills
ABSTRACT
A systematic review and meta-analysis were conducted to examine the efficacy and safety of pegylated interferon (peg-IFN) alfa-2a and peg-IFN alfa-2b plus ribavirin (RBV) in children and adolescents with chronic hepatitis C virus (HCV). Medline, Embase, and Cochrane Central Register of Controlled Trials were searched. Clinical trials examining peg-IFN alfa-2a or peg-IFN alfa-2b plus RBV among persons ages 3-18 years with HCV were included. Data were abstracted for complete early virologic response (EVR), sustained virologic response (SVR), relapse, treatment discontinuations, hematologic and dermatologic adverse events, and growth inhibition. Eight trials met the inclusion criteria. Results indicate that 70% of subjects (95% confidence interval [CI], 58%-81%) achieved EVR, and 58% (95% CI, 53%-64%) achieved SVR. EVR and SVR were higher for those with HCV genotypes 2/3 than 1/4. Discontinuation due to adverse events and discontinuation due to viral breakthrough were each 4%, discontinuation due to a lack of response was 15%, and relapse was 7%. Anemia, neutropenia, leukopenia, and thrombcytopenia were 11%, 32%, 52%, and 5%, respectively. Alopecia, injection site erythema, and pruritus were 13%, 27%, and 10%, respectively. Small growth inhibitions were observed during treatment. The results of this meta-analysis indicate that peg-IFN/RBV combination treatment is effective and safe in treating children and adolescents with HCV.
Sigma-Aldrich
Ribavirin, antiviral
Pricing and availability is not currently available.
Ribavirin, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.