Merck
  • Silica xerogels and hydroxyapatite nanocrystals for the local delivery of platinum-bisphosphonate complexes in the treatment of bone tumors: a mini-review.
Silica xerogels and hydroxyapatite nanocrystals for the local delivery of platinum-bisphosphonate complexes in the treatment of bone tumors: a mini-review.
Journal of inorganic biochemistry (2012-07-25)
Michele Iafisco, Nicola Margiotta
ABSTRACT
The present review focuses on the "drug targeting and delivery" approach of the selective transportation of cisplatin to bone tumors and bone metastases. This aim is realized by binding cisplatin to (bis)phosphonate ligands or their derivatives. Geminal bisphosphonates are in clinical use in the treatment of several bone-related diseases because of their high affinity for calcium ions and hence for bones. Platinum-bisphosphonate complexes may be easily loaded onto calcium-containing inorganic matrices, such as calcium-doped sol-gel derived silica xerogels and hydroxyapatite nanocrystals, for local administration at the site of the bone malignancy. The composites may be used as bone-filler materials that, in addition to their action as bone substitutes, can also act as controlled platinum-drug releasing agents. The release kinetics of the drug can be tailored for specific therapeutic applications modulating the physico-chemical features of the inorganic matrices. Moreover, apatite nanocrystals loaded with platinum-bisphosphonate prodrugs can be used as injectable material for nanomedical applications (e.g. intracellular drug delivery).
Sigma-Aldrich
Platinum, wire, diam. 2.0 mm, 99.9% trace metals basis
Sigma-Aldrich
Platinum, wire, diam. 0.076 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Platinum, foil, thickness 0.127 mm, 99.99% trace metals basis
Sigma-Aldrich
Platinum, foil, thickness 0.1 mm, 99.9% trace metals basis
Sigma-Aldrich
Platinum, foil, thickness 0.25 mm, 99.99% trace metals basis
Sigma-Aldrich
Platinum, wire, diam. 0.25 mm, 99.9% trace metals basis
Sigma-Aldrich
Platinum black, fuel cell grade, ≥99.9% trace metals basis
Sigma-Aldrich
Platinum, foil, thickness 0.025 mm, 99.9% trace metals basis
Sigma-Aldrich
Platinum black, black, powder, ≤20 μm, ≥99.95% trace metals basis
Sigma-Aldrich
Platinum, wire, diam. 0.127 mm, 99.9% trace metals basis
Sigma-Aldrich
Platinum, gauze, 100 mesh, 99.9% trace metals basis
Sigma-Aldrich
Platinum black, low bulk density, ≥99.9% trace metals basis
Sigma-Aldrich
Platinum, shot, ≤3 mm, ≥99.9% trace metals basis
Platinum, wire reel, 5m, diameter 0.05mm, hard, 99.99%
Platinum, wire, straight, 200mm, diameter 0.1mm, hard, 99.99%
Platinum, wire reel, 5m, diameter 0.063mm, hard, 99.9%
Platinum, foil, not light tested, 50x50mm, thickness 0.004mm, as rolled, 99.95%
Platinum, tube, 200mm, outside diameter 1.0mm, inside diameter 0.8mm, wall thickness 0.1mm, as drawn, 99.95%
Platinum, tube, 200mm, outside diameter 1.2mm, inside diameter 1.0mm, wall thickness 0.1mm, as drawn, 99.95%
Platinum, tube, 100mm, outside diameter 0.7mm, inside diameter 0.4mm, wall thickness 0.15mm, as drawn, 99.95%
Platinum, tube, 50mm, outside diameter 1.0mm, inside diameter 0.8mm, wall thickness 0.1mm, as drawn, 99.95%
Platinum, tube, 100mm, outside diameter 0.6mm, inside diameter 0.5mm, wall thickness 0.05mm, as drawn, 99.95%
Platinum, insulated wire, 0.5m, conductor diameter 0.025mm, insulation thickness 0.005mm, polyimide insulation, 99.99%
Platinum, insulated wire, 2m, conductor diameter 0.018mm, insulation thickness 0.002mm, polyester insulation, 99.99%
Platinum, tube, 100mm, outside diameter 1.0mm, inside diameter 0.8mm, wall thickness 0.1mm, as drawn, 99.95%
Platinum, rod, 10mm, diameter 4.0mm, 99.95%
Platinum, wire reel, 0.05m, diameter 0.6mm, as drawn, 99.99+%
Platinum, foil, not light tested, 50x50mm, thickness 0.0125mm, as rolled, 99.95%
Platinum, wire reel, 0.05m, diameter 1.0mm, annealed, may be used as one component of a thermocouple, 99.99%
Platinum, foil, 15x15mm, thickness 0.2mm, as rolled, 99.95%