Merck
  • Rare coding variants in the phospholipase D3 gene confer risk for Alzheimer's disease.
Rare coding variants in the phospholipase D3 gene confer risk for Alzheimer's disease.
Nature (2013-12-18)
Carlos Cruchaga, Celeste M Karch, Sheng Chih Jin, Bruno A Benitez, Yefei Cai, Rita Guerreiro, Oscar Harari, Joanne Norton, John Budde, Sarah Bertelsen, Amanda T Jeng, Breanna Cooper, Tara Skorupa, David Carrell, Denise Levitch, Simon Hsu, Jiyoon Choi, Mina Ryten, Celeste Sassi, Jose Bras, Raphael J Gibbs, Dena G Hernandez, Michelle K Lupton, John Powell, Paola Forabosco, Perry G Ridge, Christopher D Corcoran, JoAnn T Tschanz, Maria C Norton, Ronald G Munger, Cameron Schmutz, Maegan Leary, F Yesim Demirci, Mikhil N Bamne, Xingbin Wang, Oscar L Lopez, Mary Ganguli, Christopher Medway, James Turton, Jenny Lord, Anne Braae, Imelda Barber, Kristelle Brown, Pau Pastor, Oswaldo Lorenzo-Betancor, Zoran Brkanac, Erick Scott, Eric Topol, Kevin Morgan, Ekaterina Rogaeva, Andy Singleton, John Hardy, M Ilyas Kamboh, Peter St George-Hyslop, Nigel Cairns, John C Morris, John S K Kauwe, Alison M Goate
ABSTRACT
Genome-wide association studies (GWAS) have identified several risk variants for late-onset Alzheimer's disease (LOAD). These common variants have replicable but small effects on LOAD risk and generally do not have obvious functional effects. Low-frequency coding variants, not detected by GWAS, are predicted to include functional variants with larger effects on risk. To identify low-frequency coding variants with large effects on LOAD risk, we carried out whole-exome sequencing (WES) in 14 large LOAD families and follow-up analyses of the candidate variants in several large LOAD case-control data sets. A rare variant in PLD3 (phospholipase D3; Val232Met) segregated with disease status in two independent families and doubled risk for Alzheimer's disease in seven independent case-control series with a total of more than 11,000 cases and controls of European descent. Gene-based burden analyses in 4,387 cases and controls of European descent and 302 African American cases and controls, with complete sequence data for PLD3, reveal that several variants in this gene increase risk for Alzheimer's disease in both populations. PLD3 is highly expressed in brain regions that are vulnerable to Alzheimer's disease pathology, including hippocampus and cortex, and is expressed at significantly lower levels in neurons from Alzheimer's disease brains compared to control brains. Overexpression of PLD3 leads to a significant decrease in intracellular amyloid-β precursor protein (APP) and extracellular Aβ42 and Aβ40 (the 42- and 40-residue isoforms of the amyloid-β peptide), and knockdown of PLD3 leads to a significant increase in extracellular Aβ42 and Aβ40. Together, our genetic and functional data indicate that carriers of PLD3 coding variants have a twofold increased risk for LOAD and that PLD3 influences APP processing. This study provides an example of how densely affected families may help to identify rare variants with large effects on risk for disease or other complex traits.
Sigma-Aldrich
Anti-c-Myc antibody, Mouse monoclonal, clone 9E10, purified from hybridoma cell culture
Sigma-Aldrich
Phospholipase D from cabbage, Type IV, lyophilized powder, ≥100 units/mg solid
Sigma-Aldrich
Phospholipase D from Streptomyces sp., Type VII, lyophilized powder, ≥150 units/mg solid
Sigma-Aldrich
Phospholipase D from Arachis hypogaea (peanut), Type II, lyophilized powder, ≥60 units/mg protein
Sigma-Aldrich
Phospholipase D from Streptomyces chromofuscus, ≥50,000 units/mL, buffered aqueous glycerol solution