- Controlled one-step fabrication of highly oriented ZnO nanoneedle/nanorods arrays at near room temperature.
Controlled one-step fabrication of highly oriented ZnO nanoneedle/nanorods arrays at near room temperature.
Chemical communications (Cambridge, England) (2006-04-04)
Xufeng Wu, Hua Bai, Chun Li, Gewu Lu, Gaoquan Shi
PMID16583010
ABSTRACT
Highly oriented ZnO nanoneedle/nanorods arrays have been fabricated by direct oxidation of zinc foil in alkaline zincate ion solution at near room temperature (20 degrees C for nanoneedles, 30 degrees C for nanorods).
Product Description
Zinc, foil, 1m coil, thickness 0.125mm, as rolled, 99.95+%
Zinc, foil, 15mm disks, thickness 0.125mm, as rolled, 99.95+%
Zinc, foil, 10mm disks, thickness 0.25mm, as rolled, 99.99+%
Zinc, foil, 25mm disks, thickness 0.05mm, as rolled, 99.95+%
Zinc, foil, 1m coil, thickness 0.175mm, as rolled, 99.95+%
Zinc, foil, 0.3m coil, thickness 0.175mm, as rolled, 99.95+%
Zinc, foil, 15mm disks, thickness 0.005mm, 99.9%
Zinc, foil, 1m coil, thickness 0.1mm, as rolled, 99.95+%
Zinc, foil, 6mm disks, thickness 0.02mm, 99.9%
Zinc, foil, light tested, 25x25mm, thickness 0.025mm, as rolled, 99.99+%
Zinc, foil, 8mm disks, thickness 1.0mm, as rolled, 99.99+%
Zinc, foil, 10mm disks, thickness 0.0025mm, 99.9%
Zinc, foil, 10mm disks, thickness 0.02mm, 99.9%
Zinc, foil, 50x50mm, thickness 6mm, as rolled, 99.99+%
Zinc, foil, 15mm disks, thickness 1.0mm, as rolled, 99.99+%
Zinc, foil, 10mm disks, thickness 0.025mm, as rolled, 99.95+%
Zinc, foil, 4mm disks, thickness 1.0mm, as rolled, 99.95+%
Zinc, foil, 8mm disks, thickness 0.005mm, 99.9%
Zinc, foil, 25mm disks, thickness 0.20mm, as rolled, 99.95+%
Zinc, foil, not light tested, 100x100mm, thickness 0.02mm, 99.9%
Zinc, foil, light tested, 100x100mm, thickness 0.075mm, as rolled, 99.95+%
Zinc, foil, light tested, 100x100mm, thickness 0.05mm, as rolled, 99.99+%
Zinc, foil, light tested, 50x50mm, thickness 0.025mm, as rolled, 99.99+%
Zinc, foil, 25mm disks, thickness 0.25mm, as rolled, 99.95+%
Zinc, foil, 6mm disks, thickness 0.015mm, 99.9%
Zinc, foil, 8mm disks, thickness 0.25mm, as rolled, 99.95+%
Zinc, foil, 6mm disks, thickness 0.5mm, as rolled, 99.99+%
Zinc, foil, 8mm disks, thickness 0.175mm, as rolled, 99.95+%
Zinc, foil, 5m coil, thickness 0.25mm, as rolled, 99.95+%
Zinc, foil, light tested, 50x50mm, thickness 0.05mm, as rolled, 99.95+%