Merck
  • Cyclophosphamide plasma and cerebrospinal fluid kinetics with and without dimethyl sulfoxide.
Cyclophosphamide plasma and cerebrospinal fluid kinetics with and without dimethyl sulfoxide.
Clinical pharmacology and therapeutics (1982-07-01)
M J Egorin, R S Kaplan, M Salcman, J Aisner, M Colvin, P H Wiernik, N R Bachur
ABSTRACT
Ten patients with brain tumors and indwelling ventricular reservoirs were pretreated with 5% to 10% dimethyl sulfoxide (DMSO) (intravenous, oral, or both) and were then treated with 1.0 to 1.25 gm/m2 cyclophosphamide (CYC). All patients were also on anticonvulsants and dexamethasone. CYC and alkylating activity (alk act) in plasma and concomitant ventricular cerebrospinal fluid (CSF) were measured by gas chromatography and p-nitrobenzyl pyridine assay. CYC entered the CSF without difficulty and was lost from CSF more slowly than from plasma. Alk act did not enter CSF as well as did CYC. DMSO did not alter any measured aspect of CYC or alk act disposition. Specifically, it did not alter the CYC plasma half-life (t1/2), CSF t1/2, peak CSF: peak plasma CYC concentration ratio, or the urinary excretion of CYC. DMSO did not alter the plasma t1/2 or urinary excretion of alk act or the peak CSF:peak plasma concentration ratio of alk act. Our data show reduced plasma t1/2 of CYC and increased plasma and urinary alk act. This may reflect tht effect of long-term therapy with anticonvulsants or steroids.
Product Description
Supelco
Pyridine solution, certified reference material, 2000 μg/mL in methanol
Supelco
Pyridine, analytical standard
Sigma-Aldrich
Pyridine, anhydrous, 99.8%
Sigma-Aldrich
Pyridine, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Pyridine, ≥99%
Sigma-Aldrich
Pyridine, ≥99.5% (GC)
Sigma-Aldrich
Pyridine, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Pyridine, ReagentPlus®, ≥99%
Sigma-Aldrich
Pyridine, ReagentPlus®, ≥99%
Sigma-Aldrich
Pyridine, biotech. grade, ≥99.9%
Sigma-Aldrich
Pyridine, ACS reagent, ≥99.0%