Merck
Ion-interactions as driving force in polysaccharide assembly.
Carbohydrate polymers (2013-03-08)
Thomas Bechtold, Avinash P Manian, Hale B Öztürk, Uttam Paul, Barbora Široká, Ján Široký, Hossam Soliman, Loan T T Vo, Hai Vu-Manh
ABSTRACT
The structure of cellulose is characterised by extensive non-covalent interactions. Recent discussions suggest that hydrophobic interactions between polymer chains also play a significant role in governing cellulose solubility. Surprisingly in almost all cellulose shaping processes, chemical systems or solvents are applied, which base on melts or solutions of charged molecules. Ionic interactions play a significant role in the shaping and modification of cellulose based materials. Dependent on the systems used different principles govern the processes and define the results, e.g. formation of associates with alkali hydroxides, ion-exchange reactions to selectively bind multivalent ions at carboxylic groups, adsorption of dissolved polymers through surface charge neutralisation or metal complex formation, where cellulose takes the role of a polymer ligand system. Presence of charged compounds takes a significant role in cellulose swelling and dissolution, but also directs reassembly of polysaccharide material to solid three-dimensional structures.
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Sodium hydroxide, beads, 16-60 mesh, reagent grade, 97%
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, K ≤0.02%, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, reag. Ph. Eur., K ≤0.02%, ≥98%, pellets
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Sodium hydroxide solution, purum, ≥32%
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥98%, pellets
Sigma-Aldrich
Ascarite®, Sodium hydroxide-coated silica, 8-20 mesh
Sigma-Aldrich
Ascarite®, Sodium hydroxide-coated silica, 20-30 mesh