Merck
  • [Determination of beta-eudesmol in rhizome of Atractylodes lancea by RP-HPLC].
[Determination of beta-eudesmol in rhizome of Atractylodes lancea by RP-HPLC].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2008-03-04)
Yan-Ming Chen, Gui-Xin Chou, Zheng-Tao Wang
ABSTRACT
To develop a RP-HPLC method for determination of beta-eudesmol in rhizome of Atractylodes lancea, and to provide valuble data for quality control of A. lancea. The samples were separated on an Inertsil ODS-3 (4.6 mm x 250 mm, 5 microm) column with the mobile phase of acetonitrile-water (68:32). Flow rate was 1.0 mL x min(-1). The detection wavelength was set at 200 nm. Column temperature was 25 degrees C. The contents of beta-eudesmol determinated was 0.833-4.466 mg x g(-1), The linear range of beta-eudesmol was 0.048-1.200 microg (r = 0.999 9), the average recovery was 99.3%, RSD was 1.4% (n = 9). The method for quantitation of beta-eudesmol in A. lancea was accurate and reliable, which can be used to evaluate the quality of rhizome of A. lancea.
Sigma-Aldrich
β-Eudesmol, ≥90% (GC)