Merck
  • Urinary excretion measurement of cysteine and homocysteine in the form of their S-pyridinium derivatives by high-performance liquid chromatography with ultraviolet detection.
Urinary excretion measurement of cysteine and homocysteine in the form of their S-pyridinium derivatives by high-performance liquid chromatography with ultraviolet detection.
Journal of chromatography. A (1998-05-02)
E Kaniowska, G Chwatko, R Głowacki, P Kubalczyk, E Bald
ABSTRACT
Several human diseases, in particular metabolic disorders, often lead to the accumulation of characteristic metabolites in plasma, urine and cells. The selected diseases of this type include cystinuria and homocystinuria. In the typical laboratory diagnosis of these two diseases, a positive nitroprusside test is followed by quantitative analysis of urine cysteine and homocysteine in order to differentiate between cystinuria and homocystinuria. A sensitive and reproducible assay for total urine cysteine and homocysteine has been developed. The essential steps in the assay include conversion of disulphides to free thiols with tributylphosphine, conjugation of the thiols with 2-chloro-1-methyl pyridinium iodide, separation of S-pyridinium derivatives of cysteine and homocysteine from other endogenous urine thiol derivatives by reversed-phase high-performance liquid chromatography, and detection and quantitation by spectrophotometry. The method has a sensitivity of 4 pmol and is reproducible, intra- and inter-day coefficients of variation are from 1.37 to 4.14% and from 2.38 to 5.01%, respectively. The mean concentration of total urine cysteine and homocysteine in healthy donors (7 men and 7 women) were for women. 92.0 +/- 45.8 and 16.4 +/- 4.8 respectively, and for men 120.9 +/- 46.6 and 21.5 +/- 7.4 nmol/ml, respectively. Total urine homocysteine represents approximately 17.7% of cysteine in the urine of normal individuals.
Sigma-Aldrich
Tributylphosphine, mixture of isomers, 97%
Sigma-Aldrich
Tributylphosphine, ≥93.5% (Tri-N-butylphosphine, GC)
Sigma-Aldrich
Tri-n-butylphosphine, 99%
Sigma-Aldrich
Tributylphosphine solution, 200 mM (in N-methyl-2-pyrrolidinone), liquid
Sigma-Aldrich
Tri-n-butylphosphine, 97%