Merck
  • Enantiomers of cis- and trans-3-(4-propyl-cyclopent-2-enyl) propyl acetate. A study on the bioactive conformation and chiral recognition of a moth sex pheromone component.
Enantiomers of cis- and trans-3-(4-propyl-cyclopent-2-enyl) propyl acetate. A study on the bioactive conformation and chiral recognition of a moth sex pheromone component.
Bioorganic & medicinal chemistry (1998-02-12)
A L Gustavsson, M C Larsson, B S Hansson, T Liljefors
ABSTRACT
The enantiomers of cis- and trans-3-(4-propyl-cyclopent-2-enyl) propyl acetate, which are conformationally constrained analogues of (Z)-5-decenyl acetate (1), a sex pheromone component of the turnip moth, Agrotis segetum, have been synthesized and tested using the electrophysiological single-sensillum technique. The analogues mimic a cisoid and transoid conformation of 1, respectively. In addition, the enantiomers of each of the cis- and trans-isomers are conformationally constrained analogues of enantiomeric cisoid and transoid conformations of 1. Thus, the compounds prepared and tested are well suited to investigate the nature of the bioactive conformation of the natural pheromone component 1 and the chiral sense of its interaction with the receptor. Electrophysiological single-sensillum recordings show that the activity of the most active cis-isomer, which has a (1S,4R)-configuration, is more than two orders of magnitude higher than that of the most active trans-isomer. Furthermore, the (1S,4R)-isomer is at least 100 times more active than its enantiomer. These results strongly support a previously proposed cisoid bioactive conformation of 1. Furthermore, the (1S,4R)-configuration of most active stereoisomer identifies the chiral sense of the interaction between the natural pheromone component 1 and its receptor.
Sigma-Aldrich
Propyl acetate, 99%
Pricing and availability is not currently available.
Sigma-Aldrich
Propyl acetate, natural, ≥97%, FCC, FG
Pricing and availability is not currently available.
Sigma-Aldrich
Propyl acetate, ≥98%, FG
Pricing and availability is not currently available.
Supelco
Propyl acetate, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
Propyl acetate, ≥99.5%
Pricing and availability is not currently available.