Merck
  • Effect of different dosages of oral vitamin D supplementation on vitamin D status in healthy, breastfed infants: a randomized trial.
Effect of different dosages of oral vitamin D supplementation on vitamin D status in healthy, breastfed infants: a randomized trial.
JAMA (2013-05-02)
Sina Gallo, Kathryn Comeau, Catherine Vanstone, Sherry Agellon, Atul Sharma, Glenville Jones, Mary L'Abbé, Ali Khamessan, Celia Rodd, Hope Weiler
ABSTRACT
Vitamin D supplementation in infancy is required to support healthy bone mineral accretion. A supplement of 400 IU of vitamin D per day is thought to support plasma 25-hydroxyvitamin D (25[OH]D) concentrations between 40 and 50 nmol/L; some advocate 75 to 150 nmol/L for bone health. To investigate the efficacy of different dosages of vitamin D in supporting 25(OH)D concentrations in infants. Double-blind randomized clinical trial conducted among 132 one-month-old healthy, term, breastfed infants from Montréal, Québec, Canada, between March 2007 and August 2010. Infants were followed up for 11 months ending August 2011 (74% completed study). Participants were randomly assigned to receive oral cholecalciferol (vitamin D3) supplements of 400 IU/d (n=39), 800 IU/d (n=39), 1200 IU/d (n=38), or 1600 IU/d (n=16). The primary outcome was a plasma 25(OH)D concentration of 75 nmol/L or greater in 97.5% of infants at 3 months. Secondary outcomes included 25(OH)D concentrations of 75 nmol/L or greater in 97.5% of infants at 6, 9, and 12 months; 25(OH)D concentrations of 50 nmol/L or greater across all times; growth; and whole body and regional bone mineral content. Data were analyzed by intention to treat using available data, logistic regression, and mixed-model analysis of variance. By 3 months, 55% (95% CI, 38%-72%) of infants in the 400-IU/d group achieved a 25(OH)D concentration of 75 nmol/L or greater vs 81%(95% CI, 65%-91%) in the 800-IU/d group, 92% (95% CI, 77%-98%) in the 1200-IU/d group, and 100% in the 1600-IU/d group. This concentration was not sustained in 97.5% of infants at 12 months in any of the groups. The 1600-IU/d dosage was discontinued prematurely because of elevated plasma 25(OH)D concentrations. All dosages established 25(OH)D concentrations of 50 nmol/L or greater in 97% (95% CI, 94%-100%) of infants at 3 months and sustained this in 98% (95% CI, 94%-100%) to 12 months. Growth and bone mineral content did not differ by dosage. Among healthy, term, breastfed infants, only a vitamin D supplement dosage of 1600 IU/d (but not dosages of 400, 800, or 1200 IU/d) increased plasma 25(OH)D concentration to 75 nmol/L or greater in 97.5% of infants at 3 months. However, this dosage increased 25(OH)D concentrations to levels that have been associated with hypercalcemia. clinicaltrials.gov Identifier: NCT00381914.
Product Description
Supelco
Cholecalciferol (Vitamin D3), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Cholecalciferol (D3), analytical standard
Sigma-Aldrich
Cholecalciferol, analytical standard
Sigma-Aldrich
Cholecalciferol, ≥98% (HPLC)
Sigma-Aldrich
Cholecalciferol, meets USP testing specifications
Cholecalciferol, European Pharmacopoeia (EP) Reference Standard