Merck
  • A new quantitative method to determine the uptake of SPIONs in animal tissue and its application to determine the quantity of nanoparticles in the liver and lung of Balb-c mice exposed to the SPIONs.
A new quantitative method to determine the uptake of SPIONs in animal tissue and its application to determine the quantity of nanoparticles in the liver and lung of Balb-c mice exposed to the SPIONs.
Journal of biomedical nanotechnology (2013-05-01)
R D Zysler, E Lima, M Vasquez Mansilla, H E Troiani, M L Mojica Pisciotti, P Gurman, A Lamagna, L Colombo
ABSTRACT
We propose a new method for determining the quantity of superparamagnetic iron oxide nanoparticles (Fe3O4, SPIONs) embedded in animal tissue using magnetization measurements. With this method, the smallest detectable quantity of magnetite nanoparticles in a tissue sample is -1 microg. We showed that this method has proved being efficient. In this study, we focused in determining the quantity of SPION confined in lung and liver tissue of mice injected with -13 nm magnetite superparamagnetic nanoparticles. Furthermore, the method allowed us to detect the magnetite nanoparticles present in animal tissues without letting the natural iron ions present in the tissue or blood interfere with the measurements.
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr ~100,000
Supelco
Dextran, analytical standard, for GPC, 270,000
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr ~40,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, Mr ~60,000
Sigma-Aldrich
Dextran, enzymatic synth.
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr ~6,000
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr ~70,000
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 12,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 48,000-90,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 1,500,000-2,800,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 9,000-11,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 15,000-30,000
Sigma-Aldrich
Dextran solution from Leuconostoc mesenteroides, 20 % (w/w) (Autoclaved)
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 150,000
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 410,000
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 25,000
Supelco
Dextran from Leuconostoc mesenteroides, for GPC, analytical standard, Mw 50,000
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 1,000
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 670,000
Supelco
Dextran, analytical standard, for GPC, Set Mp 1,000-400,000
Supelco
Dextran, analytical standard, for GPC, 410,000
Supelco
Dextran, analytical standard, for GPC, 5,000
Supelco
Dextran, analytical standard, for GPC, 670,000
Supelco
Dextran, analytical standard, for GPC, 50,000
Supelco
Dextran, analytical standard, for GPC, 25,000
Supelco
Dextran, analytical standard, for GPC, 12,000
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr 15,000-25,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 60,000-76,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, Mr ~200,000
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr 450,000-650,000