Merck
Correlates of ecstasy use in the Swedish general population.
Substance use & misuse (2013-02-13)
Tove Abrahamsson, Anders Hakansson
ABSTRACT
The aim of the present study was to report independent correlates of ecstasy use in the Swedish general population. Data were drawn from a Swedish national household survey conducted in 2008-2009 on a random, stratified sample of 58,000 inhabitants of Sweden, aged 15 to 64. The response rate was 38.3%. Logistic regression analysis was used to identify correlates of recent (past-year) and former ecstasy use. Results are discussed and limitations of the study are noted.
Sigma-Aldrich
(±)-3,4-Methylenedioxymethamphetamine hydrochloride
Supelco
(±)-MDMA solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®