Merck
  • Reduced cytotoxicity of silver ions to mammalian cells at high concentration due to the formation of silver chloride.
Reduced cytotoxicity of silver ions to mammalian cells at high concentration due to the formation of silver chloride.
Toxicology in vitro : an international journal published in association with BIBRA (2012-12-19)
Shaokun Zhang, Chan Du, Zaizhi Wang, Xinguang Han, Kun Zhang, Lihong Liu
ABSTRACT
Silver-containing antimicrobial agents are used in various medical products. However, their toxicity to mammalian cells has not been sufficiently evaluated. Numerous studies have unveiled evidence of significant antimicrobial properties associated with Ag ions. In cell culture media or human body fluids, the free Ag(+) has rich opportunities to complex with Cl(-). Surprisingly, studies on the toxicity of solid form AgCl(s) to mammalian cells are quite limited. In this study, we evaluated the cytotoxicity of Ag ions and silver chloride colloids on red blood cells and human mesenchymal stem cells (hMSCs). The adverse effects of silver chloride on red blood cells and hMSC were viewed by SEM and LIVE/DEAD viability staining, respectively. Among different tested chemical forms of silver, AgCl was identified to be the least cytotoxic. Moreover, a decline in the cytotoxicity of AgCl at significantly high concentrations was observed. We attributed the reduced cytotoxicity to aggregated AgCl which limited the bioavailability of free Ag(+) ions.
Sigma-Aldrich
Silver chloride, ReagentPlus®, 99%
Sigma-Aldrich
Silver chloride, AnhydroBeads, −10 mesh, 99.998% trace metals basis
Sigma-Aldrich
Silver chloride, 99.999% trace metals basis