Merck
  • Biological evaluation of an ornithine-modified (99m)Tc-labeled RGD peptide as an angiogenesis imaging agent.
Biological evaluation of an ornithine-modified (99m)Tc-labeled RGD peptide as an angiogenesis imaging agent.
Nuclear medicine and biology (2012-12-15)
Irene Tsiapa, George Loudos, Alexandra Varvarigou, Eirini Fragogeorgi, Dimitrios Psimadas, Theodoros Tsotakos, Stavros Xanthopoulos, Dimitris Mihailidis, Penelope Bouziotis, George C Nikiforidis, George C Kagadis
ABSTRACT
Radiolabeled RGD peptides that specifically target integrin α(ν)β(3) have great potential in early tumor detection through noninvasive monitoring of tumor angiogenesis. Based on previous findings of our group on radiopeptides containing positively charged aminoacids, we developed a new cyclic cRGDfK derivative, c(RGDfK)-(Orn)(3)-CGG. This new peptide availing the polar linker (Orn)(3) and the (99m)Tc-chelating moiety CGG (Cys-Gly-Gly) is appropriately designed for (99m)Tc-labeling, as well as consequent conjugation onto nanoparticles. A tumor imaging agent, c(RGDfK)-(Orn)(3)-[CGG-(99m)Tc], is evaluated with regard to its radiochemical, radiobiological and imaging characteristics. The complex c(RGDfK)-(Orn)(3)-[CGG-(99m)Tc] was obtained in high radiochemical yield (>98%) and was stable in vitro and ex vivo. It presented identical to the respective, fully analytically characterized (185/187)Re complex retention time in RP-HPLC. In contrary to other RGD derivatives, we showed that the new radiopeptide exhibits kidney uptake and urine excretion due to the ornithine linker. High tumor uptake (3.87±0.48% ID/g at 60 min p.i.) was observed and was maintained relatively high even at 24 h p.i. (1.83±0.05 % ID/g), thus providing well-defined scintigraphic imaging. Accumulation in other organs was negligible. Blocking experiments indicated target specificity for integrin receptors in U87MG glioblastoma cells. Due to its relatively high tumor uptake, renal elimination and negligible abdominal localization, the new (99m)Tc-RGD peptide is considered promising in the field of imaging α(ν)β(3)-positive tumors. However, the preparation of multifunctional SPECT/MRI contrast agents (RGD-conjugated nanoparticles) for dual modality imaging of integrin expressing tumors should be further investigated.
Sigma-Aldrich
L-Ornithine monohydrochloride, BioXtra, ≥99%
Sigma-Aldrich
L-Ornithine hydrochloride, 99%
Sigma-Aldrich
D-Ornithine monohydrochloride, ~98%
Sigma-Aldrich
L-Ornithine monohydrochloride, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
L-Ornithine monohydrochloride, ≥99%
Sigma-Aldrich
L-Ornithine dihydrochloride, ≥99.0% (AT)
Sigma-Aldrich
DL-Ornithine monohydrochloride, ≥99.0% (AT)